Product Details

SNP ID
rs4924949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18953583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGGGGTGAGGTCTGCTATCAAG[A/G]GGCCCACACAGGACCAGACCTTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC5A10 PubMed Links

Gene Details

Gene
SLC5A10
Gene Name
solute carrier family 5 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042450.2 Intron NP_001035915.1
NM_001270648.1 Intron NP_001257577.1
NM_001270649.1 Intron NP_001257578.1
NM_001282417.1 Intron NP_001269346.1
NM_152351.4 Intron NP_689564.3
XM_011523652.1 Intron XP_011521954.1
XM_011523653.2 Intron XP_011521955.1
XM_017024191.1 Intron XP_016879680.1

View Full Product Details