Product Details
- SNP ID
-
rs3826231
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:406429 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCTCTCCCATGGTCCCCTGTTC[A/G]GGTGGCTGTGGGGGGGCTGGGGCTG
- Phenotype
-
MIM: 615839
MIM: 601818
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DECR2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139257963] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DECR2
- Gene Name
- 2,4-dienoyl-CoA reductase 2, peroxisomal
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020664.3 |
|
Intron |
|
|
NP_065715.1 |
- Gene
- NME4
- Gene Name
- NME/NM23 nucleoside diphosphate kinase 4
There are no transcripts associated with this gene.
View Full Product Details