Product Details

SNP ID
rs487098
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:122444760 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAAATAGTCAAACAAATTTGTTTT[C/T]ACTCTTCATTTCACAACTATAGAGC
Phenotype
MIM: 614548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERINC1 PubMed Links

Gene Details

Gene
SERINC1
Gene Name
serine incorporator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020755.3 1790 UTR 3 NP_065806.1

View Full Product Details