Product Details
- SNP ID
-
rs35681037
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:327758 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GAGAGGTAAGGTCTTTCTCTGGAAA[A/T]CCAGCAGTAGAGGAAGGGACCACCC
- Phenotype
-
MIM: 601947
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NRSN2-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45451194] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NRSN2-AS1
- Gene Name
- NRSN2 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SOX12
- Gene Name
- SRY-box 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006943.3 |
2188 |
UTR 3 |
|
|
NP_008874.2 |
View Full Product Details