Product Details
- SNP ID
-
rs1883264
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43113840 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CGGATCCCATGAGATTTTGACTACC[C/G]TCTGTGCTCAGAACAGTTTGAGCCG
- Phenotype
-
MIM: 603392
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BIK
PubMed Links
Gene Details
- Gene
- BIK
- Gene Name
- BCL2 interacting killer
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001197.4 |
|
Intron |
|
|
NP_001188.1 |
View Full Product Details