Product Details
- SNP ID
-
rs6133280
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:6013842 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTACATGGAGAGTTGCTTTTAAGCA[A/G]TGTTGAAGGAGAGTGGAGTGAAGGG
- Phenotype
-
MIM: 608188
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRLS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116255680] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRLS1
- Gene Name
- cardiolipin synthase 1
- Gene
- MCM8-AS1
- Gene Name
- MCM8 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details