Product Details

SNP ID
rs7002743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:379471 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAAGACAGCATCTGTTAAAAAAC[A/G]CAGCAGTATGTTATGTGTCACTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM87A PubMed Links

Gene Details

Gene
FAM87A
Gene Name
family with sequence similarity 87 member A
There are no transcripts associated with this gene.

View Full Product Details