Product Details

SNP ID
rs2425903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46722697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAAATGATGCGTTTAAAGTTTCT[C/T]ACACTATGCCTGGCACATAGGGATC
Phenotype
MIM: 606145
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC2A10 PubMed Links

Gene Details

Gene
SLC2A10
Gene Name
solute carrier family 2 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030777.3 Intron NP_110404.1
XM_011529060.2 Intron XP_011527362.1
XM_011529061.2 Intron XP_011527363.1
XM_011529062.2 Intron XP_011527364.1
XM_011529063.2 Intron XP_011527365.1
XM_011529064.2 Intron XP_011527366.1
XM_011529065.2 Intron XP_011527367.1
XM_017028087.1 Intron XP_016883576.1

View Full Product Details