Product Details

SNP ID
rs2838262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43329162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCACTGACCCAGGCTGGGTGG[C/T]GGCAGAGAGTGGCATTCCCACTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00322 PubMed Links

Gene Details

Gene
LINC00322
Gene Name
long intergenic non-protein coding RNA 322
There are no transcripts associated with this gene.

View Full Product Details