Product Details

SNP ID
rs523621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134225779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCCGGCCATCTGCAACTGCCAGA[C/T]GCAAAGTGAGGCTCGTCCACCGAGC
Phenotype
MIM: 609276 MIM: 610027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCAPD3 PubMed Links

Gene Details

Gene
NCAPD3
Gene Name
non-SMC condensin II complex subunit D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015261.2 Intron NP_056076.1
XM_011542719.2 Intron XP_011541021.1
Gene
VPS26B
Gene Name
VPS26, retromer complex component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052875.4 Intron NP_443107.1
XM_011542565.2 Intron XP_011540867.1

View Full Product Details