Product Details

SNP ID
rs11794994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5327154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTTTCCTAGCCAAGGGAAGCCA[C/T]AACAGACTGTAGCTGGAGAAAGGGT
Phenotype
MIM: 179730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RLN1 PubMed Links

Gene Details

Gene
RLN1
Gene Name
relaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006911.3 Intron NP_008842.1
XM_017015007.1 Intron XP_016870496.1
XM_017015008.1 Intron XP_016870497.1
XM_017015009.1 Intron XP_016870498.1
XM_017015010.1 Intron XP_016870499.1

View Full Product Details