Product Details

SNP ID
rs11128623
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:12898330 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAACAATGTATCTTCCACAAACTTT[C/T]GATAAGCTTGCAAATAAAACAGAAT
Phenotype
MIM: 610166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQSEC1 PubMed Links

Gene Details

Gene
IQSEC1
Gene Name
IQ motif and Sec7 domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134382.2 6439 UTR 3 NP_001127854.1
NM_014869.6 6439 UTR 3 NP_055684.3
XM_011534304.2 6439 UTR 3 XP_011532606.1
XM_011534305.2 6439 UTR 3 XP_011532607.1
XM_011534306.2 6439 UTR 3 XP_011532608.1
XM_011534307.2 6439 UTR 3 XP_011532609.1
XM_011534308.2 6439 UTR 3 XP_011532610.1
XM_011534310.2 6439 UTR 3 XP_011532612.1
XM_011534311.2 6439 UTR 3 XP_011532613.1
XM_011534312.2 6439 UTR 3 XP_011532614.1
XM_011534313.2 6439 UTR 3 XP_011532615.1
XM_011534314.2 6439 UTR 3 XP_011532616.1
XM_011534315.2 6439 UTR 3 XP_011532617.1
XM_017007581.1 6439 UTR 3 XP_016863070.1
XM_017007582.1 6439 UTR 3 XP_016863071.1

View Full Product Details