Product Details

SNP ID
rs9809596
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:12799014 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCTGGCTTTATGGGCAGGAGGTGCC[C/G]CATTACCACGGAGGCTCCAGCAGTC
Phenotype
MIM: 610403
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAND2 PubMed Links

Gene Details

Gene
CAND2
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162499.1 Intron NP_001155971.1
NM_012298.2 Intron NP_036430.1
XM_011533503.2 Intron XP_011531805.1
XM_011533504.2 Intron XP_011531806.1

View Full Product Details