Product Details
- SNP ID
-
rs2648535
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9366759 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGATTAGTAACCAAAGAGATGAGA[A/C]ACTAGGGAGATGGGACATTTTAAAA
- Phenotype
-
MIM: 606525
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SRGAP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74279379] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SRGAP3
- Gene Name
- SLIT-ROBO Rho GTPase activating protein 3
There are no transcripts associated with this gene.
- Gene
- THUMPD3
- Gene Name
- THUMP domain containing 3
View Full Product Details