Product Details

SNP ID
rs35969491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:11186414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTTAAAATAGAGAGTTTATCA[A/T]AGAGAGTTTTACTTTCATCTAAATA
Phenotype
MIM: 613966
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R42 PubMed Links

Gene Details

Gene
TAS2R42
Gene Name
taste 2 receptor member 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181429.2 524 Missense Mutation TAT,TTT Y175F NP_852094.2

View Full Product Details