Product Details

SNP ID
rs12403273
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:156218317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGATGATGTCCTTTGAGGAACAG[A/T]AGTCTTCAATTAAATAAAAAGCAGC
Phenotype
MIM: 609176 MIM: 610824
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PMF1 PubMed Links

Gene Details

Gene
PMF1
Gene Name
polyamine-modulated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199653.1 Intron NP_001186582.1
NM_001199654.1 Intron NP_001186583.1
NM_007221.3 Intron NP_009152.2
Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199661.1 Intron NP_001186590.1
NM_001199662.1 Intron NP_001186591.1
NM_001199663.1 Intron NP_001186592.1
NM_001199664.1 Intron NP_001186593.1
Gene
SLC25A44
Gene Name
solute carrier family 25 member 44
There are no transcripts associated with this gene.

View Full Product Details