Product Details
- SNP ID
-
rs861998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
38
- Location
-
Chr.1:157574088 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATTTGTAAAGATCAAATCAGCGTA[A/G]TTGGGATATCTGTCACCTTAAATAT
- Phenotype
-
MIM: 605876
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FCRL4
PubMed Links
Gene Details
- Gene
- FCRL4
- Gene Name
- Fc receptor like 4
View Full Product Details