Product Details

SNP ID
rs7927406
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:124673907 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAGAAAAAACGGTTACCCAGCAAC[G/T]AGAAAAACAACCGGAACCGGCGGCA
Phenotype
MIM: 610079 MIM: 608621
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIAE PubMed Links

Gene Details

Gene
SIAE
Gene Name
sialic acid acetylesterase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199922.1 64 Intron NP_001186851.1
NM_170601.4 64 Intron NP_733746.1
XM_017017930.1 64 Intron XP_016873419.1
Gene
SPA17
Gene Name
sperm autoantigenic protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017425.3 64 UTR 5 NP_059121.1
XM_011542870.2 64 Intron XP_011541172.1

View Full Product Details