Product Details

SNP ID
rs1378528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9709484 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCTTGTGCGAACTGATCTGTCAA[G/T]ATGGGGGTACAAGAACTGTAGAAAG
Phenotype
MIM: 605694
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB31 PubMed Links

Gene Details

Gene
RAB31
Gene Name
RAB31, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006868.3 Intron NP_006859.2
XM_017025528.1 Intron XP_016881017.1
XM_017025529.1 Intron XP_016881018.1

View Full Product Details