Product Details

SNP ID
rs34671397
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134071650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTCTTCAGATAAGACAACTT[A/G]TGTGCTGAAGCATATCAAAATCTGA
Phenotype
MIM: 606871
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
JAM3 PubMed Links
Additional Information
For this assay, SNP(s) [rs75604138] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
JAM3
Gene Name
junctional adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205329.1 Intron NP_001192258.1
NM_032801.4 Intron NP_116190.3

View Full Product Details