Product Details

SNP ID
rs35035485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151364594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGAAAAGGAGGACAGGAAGGTC[C/G]TGAGATAGAAGCAGAAAGAAACTGG
Phenotype
MIM: 600410
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929319 PubMed Links

Gene Details

Gene
LOC101929319
Gene Name
uncharacterized LOC101929319
There are no transcripts associated with this gene.

Gene
MIR4773-1
Gene Name
microRNA 4773-1
There are no transcripts associated with this gene.

Gene
MIR4773-2
Gene Name
microRNA 4773-2
There are no transcripts associated with this gene.

Gene
TNFAIP6
Gene Name
TNF alpha induced protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007115.3 Intron NP_009046.2

View Full Product Details