Product Details
- SNP ID
-
rs11620680
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:64550391 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCAGCCCCGCTCTGGATAGAAAGGC[A/T]GGTGGGTGCTAGCTTCCACGGTGAA
- Phenotype
-
MIM: 140560
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HSPA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114080267] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSPA2
- Gene Name
- heat shock protein family A (Hsp70) member 2
There are no transcripts associated with this gene.
- Gene
- PPP1R36
- Gene Name
- protein phosphatase 1 regulatory subunit 36
View Full Product Details