Product Details

SNP ID
rs8016222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64554969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAAAGGCAAGTGCAAAATTAG[A/G]GTGGGAAGTGGAATGATTGAGGCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R36 PubMed Links

Gene Details

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172365.2 Intron NP_758953.1
XM_005267354.3 Intron XP_005267411.1
XM_005267355.3 Intron XP_005267412.1
XM_011536468.2 Intron XP_011534770.1

View Full Product Details