Product Details

SNP ID
rs12654962
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:134971922 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAGCAGAAGAATGAGCTCAGTGA[C/G]GTTAGAACCACACCATCTTCTAAAA
Phenotype
MIM: 609120 MIM: 609836
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CATSPER3 PubMed Links

Gene Details

Gene
CATSPER3
Gene Name
cation channel sperm associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178019.2 Intron NP_821138.1
Gene
PCBD2
Gene Name
pterin-4 alpha-carbinolamine dehydratase 2
There are no transcripts associated with this gene.

View Full Product Details