Product Details

SNP ID
rs17518673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134917343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCTGTCCCTGTTCTTTCACTT[C/T]TGAATACCTGGGGCTACCCTTGTTC
Phenotype
MIM: 609836
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCBD2 PubMed Links

Gene Details

Gene
PCBD2
Gene Name
pterin-4 alpha-carbinolamine dehydratase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032151.4 Intron NP_115527.3

View Full Product Details