Product Details

SNP ID
rs4684304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCTCTCTGAGGAGGATTCCAG[A/G]CCCCCCCGTAGATCTCCGCCCTCTC
Phenotype
MIM: 607416
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHL1 PubMed Links

Gene Details

Gene
CHL1
Gene Name
cell adhesion molecule L1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253387.1 Intron NP_001240316.1
NM_001253388.1 Intron NP_001240317.1
NM_006614.3 Intron NP_006605.2
XM_006712938.1 Intron XP_006713001.1
XM_006712939.3 Intron XP_006713002.1
XM_006712940.3 Intron XP_006713003.1
XM_011533292.1 Intron XP_011531594.1
XM_011533294.1 Intron XP_011531596.1
XM_011533295.1 Intron XP_011531597.1
XM_011533296.1 Intron XP_011531598.1
XM_017005566.1 Intron XP_016861055.1
XM_017005567.1 Intron XP_016861056.1
XM_017005568.1 Intron XP_016861057.1
XM_017005569.1 Intron XP_016861058.1
XM_017005570.1 Intron XP_016861059.1
XM_017005571.1 Intron XP_016861060.1
XM_017005572.1 Intron XP_016861061.1
XM_017005573.1 Intron XP_016861062.1
Gene
LOC101927174
Gene Name
uncharacterized LOC101927174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017007606.1 Intron XP_016863095.1

View Full Product Details