Product Details

SNP ID
rs7897619
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:89330794 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCTTAATTCACCACATTAGCTCCA[C/T]AAATAAAGGGGAAAACTGCTTTTTA
Phenotype
MIM: 604650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIT3 PubMed Links

Gene Details

Gene
IFIT3
Gene Name
interferon induced protein with tetratricopeptide repeats 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031683.3 Intron NP_001026853.1
NM_001289758.1 Intron NP_001276687.1
NM_001289759.1 Intron NP_001276688.1
NM_001549.5 Intron NP_001540.2

View Full Product Details