Product Details

SNP ID
rs229361
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:20743947 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCATTCTGTCTTTATTAAGCATGA[C/G]CTATTGAAAGTTCTGTTTACGTTGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00320 PubMed Links

Gene Details

Gene
LINC00320
Gene Name
long intergenic non-protein coding RNA 320
There are no transcripts associated with this gene.

View Full Product Details