Product Details

SNP ID
rs229366
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:20746301 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAAATATCGTAATTTAGATGTGATG[C/T]AACATTTATCTTTGCAAGGAAAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00320 PubMed Links

Gene Details

Gene
LINC00320
Gene Name
long intergenic non-protein coding RNA 320
There are no transcripts associated with this gene.

View Full Product Details