Product Details

SNP ID
rs35672130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:26567156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGGAGGATCAAACTGACATGG[G/T]TTATTATATATTTTCTAGTTGTGAG
Phenotype
MIM: 605068
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WASF3 PubMed Links

Gene Details

Gene
WASF3
Gene Name
WAS protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291965.1 Intron NP_001278894.1
NM_006646.5 Intron NP_006637.2
XM_011534889.1 Intron XP_011533191.1
XM_011534890.1 Intron XP_011533192.1

View Full Product Details