Product Details

SNP ID
rs4678953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37247743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTCTCATCTGGAGCTCAGGATC[C/T]TTTTGTATGCCCATGTGGTTGTGGT
Phenotype
MIM: 602509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGA4 PubMed Links

Gene Details

Gene
GOLGA4
Gene Name
golgin A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172713.1 Intron NP_001166184.1
NM_002078.4 Intron NP_002069.2
XM_005265069.3 Intron XP_005265126.1
XM_005265070.3 Intron XP_005265127.1
XM_005265071.3 Intron XP_005265128.1
XM_005265072.3 Intron XP_005265129.1
XM_005265073.1 Intron XP_005265130.1
XM_005265074.1 Intron XP_005265131.1
XM_005265075.1 Intron XP_005265132.1
XM_006713110.2 Intron XP_006713173.1
XM_011533598.2 Intron XP_011531900.1
XM_017006184.1 Intron XP_016861673.1
XM_017006185.1 Intron XP_016861674.1
XM_017006186.1 Intron XP_016861675.1
XM_017006187.1 Intron XP_016861676.1
XM_017006188.1 Intron XP_016861677.1
Gene
LOC152048
Gene Name
uncharacterized FLJ31715
There are no transcripts associated with this gene.

View Full Product Details