Product Details

SNP ID
rs11642334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:368429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCAAATTCCTAGGCAGGCAGAG[A/G]TGGAAAGGGCAGTGAGGCCCAGGGC
Phenotype
MIM: 604853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL28 PubMed Links

Gene Details

Gene
MRPL28
Gene Name
mitochondrial ribosomal protein L28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006428.4 Intron NP_006419.2
XM_005255041.1 Intron XP_005255098.1
XM_011522351.2 Intron XP_011520653.1
Gene
TMEM8A
Gene Name
transmembrane protein 8A
There are no transcripts associated with this gene.

View Full Product Details