Product Details

SNP ID
rs2659117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50887178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTGTAGGGCACTGGGAAGGGTG[A/G]CCACATAGGAAGAAGTCTCTGTTCT
Phenotype
MIM: 147960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK2 PubMed Links

Gene Details

Gene
KLK2
Gene Name
kallikrein related peptidase 2
There are no transcripts associated with this gene.

Gene
KLKP1
Gene Name
kallikrein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details