Product Details

SNP ID
rs9646148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22830886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCATACAGCTAGGTAGTAGCAAA[G/T]CCAGGACTAGAACATAGGCCAGTCC
Phenotype
MIM: 600754 MIM: 611856
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MMP14 PubMed Links

Gene Details

Gene
MMP14
Gene Name
matrix metallopeptidase 14
There are no transcripts associated with this gene.

Gene
MRPL52
Gene Name
mitochondrial ribosomal protein L52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318870.1 Intron NP_001305799.1
NM_178336.2 Intron NP_848026.1
NM_180982.2 Intron NP_851313.1
NM_181304.2 Intron NP_851821.1
NM_181305.2 Intron NP_851822.1
NM_181306.2 Intron NP_851823.1
NM_181307.2 Intron NP_851824.1
XM_005267325.4 Intron XP_005267382.1
XM_005267328.3 Intron XP_005267385.1

View Full Product Details