Product Details

SNP ID
rs342903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146584509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTATGACTGGGAGAAAGGTCGCA[C/T]GTGATTTTAAAATATAGTTATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLSCR5 PubMed Links

Gene Details

Gene
PLSCR5
Gene Name
phospholipid scramblase family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085420.1 Intron NP_001078889.1
NM_001321245.1 Intron NP_001308174.1
XM_017006373.1 Intron XP_016861862.1

View Full Product Details