Product Details
- SNP ID
-
rs415580
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52470456 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAATAATGGCTAATGACTGCCTGAT[G/T]GGTCTAGCAAAAAATGATGCTTCTT
- Phenotype
-
MIM: 612315
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6C
PubMed Links
Gene Details
- Gene
- KRT6C
- Gene Name
- keratin 6C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173086.4 |
|
Intron |
|
|
NP_775109.2 |
View Full Product Details