Product Details
- SNP ID
-
rs7535923
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:17756290 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAGTGGGGGACTTATCTTTGGGGA[A/T]CCTATGAAAGCCCCCAGGGGCTCTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ACTL8
PubMed Links
Gene Details
- Gene
- ACTL8
- Gene Name
- actin like 8
View Full Product Details