Product Details

SNP ID
rs3804278
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:41337716 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAGAAGAAATTTTAAAGCAATTCC[G/T]ACAAGCTCAACAGTTTTATATTTAA
Phenotype
MIM: 604531
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NCR2 PubMed Links

Gene Details

Gene
NCR2
Gene Name
natural cytotoxicity triggering receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199509.1 Intron NP_001186438.1
NM_001199510.1 Intron NP_001186439.1
NM_004828.3 Intron NP_004819.2
XM_017011500.1 Intron XP_016866989.1

View Full Product Details