Product Details

SNP ID
rs11070408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43742590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTGTCACTAAAAAATAAAGTGG[A/G]AGACTTCTCTACAACAGTGAGAATG
Phenotype
MIM: 602046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPER2P1 PubMed Links

Gene Details

Gene
CATSPER2P1
Gene Name
cation channel sperm associated 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
PDIA3
Gene Name
protein disulfide isomerase family A member 3
There are no transcripts associated with this gene.

View Full Product Details