Product Details

SNP ID
rs1941030
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:60458285 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATGCAAAACTATTTTGAGTGGAA[A/G]AGAATGAAGATGAGGTAACAACAGT
Phenotype
MIM: 112210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A1 PubMed Links

Gene Details

Gene
MS4A1
Gene Name
membrane spanning 4-domains A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021950.3 Intron NP_068769.2
NM_152866.2 Intron NP_690605.1

View Full Product Details