Product Details

SNP ID
rs274712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6723837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCGCAGTAGTACGCTCAGGTTC[A/G]TGCTTTCTTGAGGTGTTCCAGAACT
Phenotype
MIM: 605198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAPD7 PubMed Links

Gene Details

Gene
PAPD7
Gene Name
PAP associated domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171805.1 Intron NP_001165276.1
NM_001171806.1 Intron NP_001165277.1
NM_006999.4 Intron NP_008930.1
XM_005248234.3 Intron XP_005248291.1
XM_011513945.1 Intron XP_011512247.1
XM_017008988.1 Intron XP_016864477.1
XM_017008989.1 Intron XP_016864478.1
XM_017008990.1 Intron XP_016864479.1

View Full Product Details