Product Details

SNP ID
rs12907665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40979243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGGCAGGGACAGGCAGAAACA[C/T]CTCCCATGCAAACACTGCCCCTCTG
Phenotype
MIM: 610169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INO80 PubMed Links

Gene Details

Gene
INO80
Gene Name
INO80 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017553.2 3779 UTR 3 NP_060023.1
XM_011521685.2 3779 Intron XP_011519987.1
XM_011521686.2 3779 UTR 3 XP_011519988.1

View Full Product Details