Product Details

SNP ID
rs412523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:4716515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCATGGAGGTTAAATAGCTTGCC[A/T]AGGGCAAAAGAGACGGTAATTTTCA
Phenotype
MIM: 609290 MIM: 610346
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AK3 PubMed Links

Gene Details

Gene
AK3
Gene Name
adenylate kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199852.1 Intron NP_001186781.1
NM_001199853.1 Intron NP_001186782.1
NM_001199855.1 Intron NP_001186784.1
NM_001199856.1 Intron NP_001186785.1
NM_016282.3 Intron NP_057366.2
Gene
CDC37L1
Gene Name
cell division cycle 37 like 1
There are no transcripts associated with this gene.

View Full Product Details