Product Details
- SNP ID
-
rs2607415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40756782 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGACATGGGAAGAAATTTAGACGT[C/G]GTGGTCAGAGGTGGCAACATTTCCT
- Phenotype
-
MIM: 182285
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C19orf54
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115527690] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C19orf54
- Gene Name
- chromosome 19 open reading frame 54
There are no transcripts associated with this gene.
- Gene
- SNRPA
- Gene Name
- small nuclear ribonucleoprotein polypeptide A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004596.4 |
|
Intron |
|
|
NP_004587.1 |
View Full Product Details