Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014203.2 | Intron | NP_055018.2 | |||
NM_130787.2 | Intron | NP_570603.2 | |||
XM_011526556.2 | Intron | XP_011524858.1 | |||
XM_011526557.2 | Intron | XP_011524859.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171937.1 | Intron | NP_001165408.1 | |||
NM_025129.4 | Intron | NP_079405.2 | |||
XM_006723399.3 | Intron | XP_006723462.1 | |||
XM_011527339.1 | Intron | XP_011525641.1 | |||
XM_011527340.1 | Intron | XP_011525642.1 | |||
XM_011527341.2 | Intron | XP_011525643.1 | |||
XM_011527342.1 | Intron | XP_011525644.1 | |||
XM_011527343.1 | Intron | XP_011525645.1 | |||
XM_011527345.1 | Intron | XP_011525647.1 | |||
XM_011527346.1 | Intron | XP_011525648.1 | |||
XM_011527347.1 | Intron | XP_011525649.1 | |||
XM_017027319.1 | Intron | XP_016882808.1 | |||
XM_017027320.1 | Intron | XP_016882809.1 | |||
XM_017027321.1 | Intron | XP_016882810.1 | |||
XM_017027322.1 | Intron | XP_016882811.1 | |||
XM_017027323.1 | Intron | XP_016882812.1 |