Product Details

SNP ID
rs10421463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50050718 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTGATGAATGTCACTAGACAGT[C/G]GGGCCCTCAAGGACCTGGCAAATGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLJ26850 PubMed Links

Gene Details

Gene
FLJ26850
Gene Name
FLJ26850 protein
There are no transcripts associated with this gene.

Gene
LOC100287477
Gene Name
zinc finger protein 420-like
There are no transcripts associated with this gene.

Gene
ZNF473
Gene Name
zinc finger protein 473
There are no transcripts associated with this gene.

View Full Product Details