Product Details
- SNP ID
-
rs28485712
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:210483082 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGTTTAATTTGATCTTCTTCAGG[C/T]GGAATAATTCAATTTTATACCCTTT
- Phenotype
-
MIM: 608307
MIM: 604155
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79894857] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPS1
- Gene Name
- carbamoyl-phosphate synthase 1
- Gene
- LANCL1
- Gene Name
- LanC like 1
There are no transcripts associated with this gene.
View Full Product Details