Product Details

SNP ID
rs11841349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32404291 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATACCTGAAAATAGCAGGATACA[A/T]AATGAGGAACTAGAAAAACAAAGAA
Phenotype
MIM: 600185
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BRCA2 PubMed Links

Gene Details

Gene
BRCA2
Gene Name
BRCA2, DNA repair associated
There are no transcripts associated with this gene.

Gene
N4BP2L1
Gene Name
NEDD4 binding protein 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079691.1 Intron NP_001073159.1
NM_001286459.1 Intron NP_001273388.1
NM_001286460.1 Intron NP_001273389.1
NM_001286461.1 Intron NP_001273390.1
NM_052818.2 Intron NP_438169.2
XM_006719890.3 Intron XP_006719953.1
XM_011535301.2 Intron XP_011533603.1
XM_011535302.2 Intron XP_011533604.1
XM_011535303.2 Intron XP_011533605.1
XM_011535304.2 Intron XP_011533606.1
XM_017020836.1 Intron XP_016876325.1
XM_017020837.1 Intron XP_016876326.1
XM_017020838.1 Intron XP_016876327.1
XM_017020839.1 Intron XP_016876328.1
XM_017020840.1 Intron XP_016876329.1
XM_017020841.1 Intron XP_016876330.1
XM_017020842.1 Intron XP_016876331.1

View Full Product Details