Product Details

SNP ID
rs3828760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:81747267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAACTTTGAAACAACTAAGCAGG[C/T]GGAGCTCTGGTTTCTGATAGGTGAG
Phenotype
MIM: 611357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM46A PubMed Links

Gene Details

Gene
FAM46A
Gene Name
family with sequence similarity 46 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017633.2 4075 UTR 3 NP_060103.2

View Full Product Details