Product Details

SNP ID
rs279887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:979080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCCAGGACTAGGGCAGAGGC[A/C]TGAGTTAACATCAGAGGTTAACTCA
Phenotype
MIM: 602424 MIM: 614754
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMRT1 PubMed Links

Gene Details

Gene
DMRT1
Gene Name
doublesex and mab-3 related transcription factor 1
There are no transcripts associated with this gene.

Gene
DMRT3
Gene Name
doublesex and mab-3 related transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021240.3 Intron NP_067063.1

View Full Product Details